Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289034.1 | NP_001275963.1 | 5 | 965 | ||
NM_001289035.1 | NP_001275964.1 | 4 | 500 | |||
NM_138343.3 | NP_612352.1 | 5 | 731 | |||
NM_201521.2 | NP_958929.1 | 5 | 731 | |||
NM_201522.2 | NP_958930.1 | 6 | 1147 | |||
NM_201523.2 | NP_958931.1 | 5 | 1021 | |||
XM_011514989.1 | XP_011513291.1 | 5 | 1080 | |||
XM_017011457.1 | XP_016866946.1 | 5 | 731 | |||
GenBank mRNA | AK055293.1 | - | 6 | 1109 | ||
AK056393.1 | - | 5 | 691 | |||
AK094102.1 | - | 5 | 987 | |||
AK127018.1 | - | 8 | 1996 | |||
AK127894.1 | - | 5 | 704 | |||
AK297433.1 | - | 4 | 463 | |||
BC012357.1 | - | 5 | 672 | |||
BC080637.1 | - | 5 | 671 | 6573021 | ||
BC103727.1 | - | 5 | 669 | 6157949 | ||
BX640717.1 | - | 5 | 928 |