Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289034.1 | NP_001275963.1 | 3 | 821 | ||
NM_138343.3 | NP_612352.1 | 3 | 587 | |||
NM_201521.2 | NP_958929.1 | 3 | 587 | |||
NM_201522.2 | NP_958930.1 | 4 | 1003 | |||
NM_201523.2 | NP_958931.1 | 3 | 877 | |||
XM_011514989.1 | XP_011513291.1 | 3 | 936 | |||
XM_017011457.1 | XP_016866946.1 | 3 | 587 | |||
GenBank mRNA | AK055293.1 | - | 4 | 965 | ||
AK056393.1 | - | 3 | 547 | |||
AK094102.1 | - | 3 | 843 | |||
AK127018.1 | - | 6 | 1852 | |||
AK127894.1 | - | 3 | 560 | |||
BC012357.1 | - | 3 | 528 | |||
BC080637.1 | - | 3 | 527 | 6573021 | ||
BC103727.1 | - | 3 | 525 | 6157949 | ||
BQ688778.1 | - | 3 | 657 | |||
BX640717.1 | - | 3 | 784 |