Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305163.1 | NP_001292092.1 | 2, 3 | 1265 | ||
NM_001305164.1 | NP_001292093.1 | 2, 3 | 644 | |||
NM_133368.2 | NP_588609.1 | 2, 3 | 666 | |||
XM_005256220.1 | XP_005256277.1 | 2, 3 | 973 | |||
GenBank mRNA | AB075852.1 | - | 2, 3 | 586 | ||
AK027549.1 | - | 2, 3 | 1216 | |||
AK172845.1 | - | 2, 3 | 622 | |||
BC013173.2 | - | 2, 3 | 621 | 4340287 |