Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305163.1 | NP_001292092.1 | 10 | 2008 | ||
NM_001305164.1 | NP_001292093.1 | 10 | 1387 | |||
NM_133368.2 | NP_588609.1 | 10 | 1409 | |||
XM_005256220.1 | XP_005256277.1 | 10 | 1716 | |||
XM_011523427.1 | XP_011521729.1 | 11 | 1495 | |||
XM_011523428.1 | XP_011521730.1 | 11 | 1809 | |||
XM_011523430.1 | XP_011521732.1 | 11 | 1436 | |||
XM_017023844.1 | XP_016879333.1 | 11 | 2182 | |||
GenBank mRNA | AB075852.1 | - | 10 | 1329 | ||
AK027549.1 | - | 10 | 1959 | |||
AK124601.1 | - | 2 | 2326 | |||
AK172845.1 | - | 10 | 1365 | |||
BC013173.2 | - | 10 | 1364 | 4340287 |