Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163321.2 | NP_001156793.2 | 10 | 1900 | ||
NM_001163322.2 | NP_001156794.1 | 10 | 1841 | |||
NM_001163323.2 | NP_001156795.1 | 10 | 1877 | |||
NM_001271835.1 | NP_001258764.1 | 10 | 1877 | |||
NM_001271836.1 | NP_001258765.1 | 10 | 2229 | |||
NM_033626.3 | NP_296375.1 | 10 | 1818 | |||
GenBank mRNA | AK056290.1 | - | 1 | 414 | ||
AK125424.1 | - | 1 | 1412 | |||
AK291370.1 | - | 10 | 2089 | |||
AK293898.1 | - | 10 | 1786 | |||
AK294024.1 | - | 10 | 1864 | |||
AK300366.1 | - | 10 | 1828 | |||
AK315837.1 | - | 10 | 1844 | |||
BC008769.2 | - | 10 | 1799 | 3139519 |