Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306193.1 | NP_001293122.1 | 12 | 1552 | ||
NM_194277.2 | NP_919253.1 | 12 | 1599 | |||
XM_017029947.1 | XP_016885436.1 | 12 | 1778 | |||
XM_017029948.1 | XP_016885437.1 | 9 | 1240 | |||
XM_017029949.1 | XP_016885438.1 | 8 | 1561 | |||
GenBank mRNA | AK125336.1 | - | 12 | 1599 | ||
AL161984.1 | - | 8 | 1260 | |||
BC114371.1 | - | 12 | 1566 | 40079764 | ||
FJ717411.1 | - | 12 | 1376 |