Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 599 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 617 | |||
NM_033421.3 | NP_219489.1 | 4 | 606 | |||
NM_152897.2 | NP_690857.1 | 4 | 606 | |||
XM_005260608.4 | XP_005260665.1 | 2 | 681 | |||
XM_011529095.2 | XP_011527397.1 | 2 | 585 | |||
XM_011529098.2 | XP_011527400.1 | 5 | 1119 | |||
GenBank mRNA | AF258570.1 | - | 1 | 266 | ||
AF395845.1 | - | 4 | 517 | |||
AF523834.1 | - | 4 | 585 | |||
AK091359.1 | - | 3 | 621 | |||
AK095851.1 | - | 3 | 480 | |||
AK226120.1 | - | 3 | 130 | |||
AK293443.1 | - | 5 | 618 | |||
AK297622.1 | - | 4 | 540 | |||
AL591562.1 | - | 4 | 585 | |||
BC012904.1 | - | 6 | 837 | |||
BC019823.1 | - | 4 | 579 | 4453502 | ||
BM925584.1 | - | 5 | 603 | |||
KU178820.1 | - | 4 | 517 | |||
KU178821.1 | - | 5 | 528 |