Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138349.2 | NP_612358.3 | 7 | 1427 | ||
XM_017025282.1 | XP_016880771.1 | 7 | 1799 | |||
GenBank mRNA | AB110209.1 | - | 7 | 1494 | ||
AK075341.1 | - | 7 | 1458 | |||
AK093030.1 | - | 1 | 2443 | |||
AL137404.1 | - | 1 | 802 | |||
BC001593.2 | - | 7 | 1427 | 2988166 | ||
BC004507.2 | - | 2 | 799 |