Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195752.1 | NP_001182681.1 | 1, 2 | 142 | ||
NM_001195753.1 | NP_001182682.1 | 2, 3 | 232 | |||
NM_138350.3 | NP_612359.2 | 1, 2 | 142 | |||
XM_005263532.3 | XP_005263589.1 | 2, 3 | 452 | |||
XM_011542401.2 | XP_011540703.1 | 1, 2 | 100 | |||
XM_017002761.1 | XP_016858250.1 | 1, 2 | 100 | |||
GenBank mRNA | AK096217.1 | - | 1, 2 | 100 | ||
BC092427.1 | - | 2, 3 | 232 | 30376814 | ||
BE905510.1 | - | 201 |