Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 611 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 729 | |||
NR_102299.1 | 6 | 984 | ||||
NR_102300.1 | 6 | 965 | ||||
NR_102301.1 | 5 | 866 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 826 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 944 | |||
GenBank mRNA | AK289841.1 | - | 5 | 720 | ||
AK292449.1 | - | 4 | 580 | |||
AY442330.1 | - | 4 | 612 | |||
AY442331.1 | - | 5 | 730 | |||
CR936599.1 | - | 4 | 577 |