Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 799 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 917 | |||
NR_102299.1 | 6 | 1172 | ||||
NR_102300.1 | 6 | 1153 | ||||
NR_102301.1 | 5 | 1054 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 1014 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 1132 | |||
GenBank mRNA | AK289841.1 | - | 5 | 908 | ||
AK292449.1 | - | 4 | 768 | |||
AY442330.1 | - | 4 | 800 | |||
AY442331.1 | - | 5 | 918 | |||
CR936599.1 | - | 4 | 765 | |||
CR936695.1 | - | 4 | 769 |