Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102603.1 | NP_001096073.1 | 7 | 2056 | ||
NM_001322128.1 | NP_001309057.1 | 7 | 2064 | |||
NM_001322129.1 | NP_001309058.1 | 6 | 2159 | |||
NM_001322130.1 | NP_001309059.1 | 7 | 2288 | |||
NM_001322131.1 | NP_001309060.1 | 6 | 2163 | |||
NM_001322132.1 | NP_001309061.1 | 7 | 2052 | |||
NM_001322133.1 | NP_001309062.1 | 7 | 2076 | |||
NM_001322134.1 | NP_001309063.1 | 7 | 2080 | |||
NM_001322135.1 | NP_001309064.1 | 6 | 1935 | |||
NM_001322136.1 | NP_001309065.1 | 5 | 1792 | |||
NM_001322137.1 | NP_001309066.1 | 8 | 2112 | |||
NM_001322138.1 | NP_001309067.1 | 7 | 1995 | |||
NM_001322139.1 | NP_001309068.1 | 9 | 2209 | |||
NM_033288.3 | NP_150630.2 | 6 | 1939 | |||
NM_198893.2 | NP_942596.1 | 7 | 2024 | |||
XM_017027445.1 | XP_016882934.1 | 4 | 2288 | |||
XM_017027446.1 | XP_016882935.1 | 6 | 2249 | |||
GenBank mRNA | AB046831.1 | - | 3 | 2409 | ||
AF277623.1 | - | 6 | 1947 | |||
AK024442.1 | - | 1 | 1303 | |||
BC094880.1 | - | 7 | 1941 | 3066532 | ||
CR749613.1 | - | 1 | 754 |