Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293083.1 | NP_001280012.1 | 47 | 5343 | ||
XM_011512110.2 | XP_011510412.1 | 47 | 5387 | |||
XM_011512111.2 | XP_011510413.1 | 47 | 5384 | |||
XM_011512113.2 | XP_011510415.1 | 47 | 5381 | |||
XM_011512114.2 | XP_011510416.1 | 47 | 5357 | |||
XM_011512115.2 | XP_011510417.1 | 46 | 5318 | |||
XM_011512116.2 | XP_011510418.1 | 47 | 5300 | |||
XM_011512117.2 | XP_011510419.1 | 46 | 5282 | |||
XM_011512118.2 | XP_011510420.1 | 46 | 5267 | |||
XM_011512119.2 | XP_011510421.1 | 46 | 5225 | |||
XM_011512120.2 | XP_011510422.1 | 46 | 5278 | |||
XM_011512122.2 | XP_011510424.1 | 47 | 5388 | |||
XM_011512124.2 | XP_011510426.1 | 31 | 4971 | |||
XM_017005222.1 | XP_016860711.1 | 47 | 5384 | |||
XM_017005223.1 | XP_016860712.1 | 30 | 4035 | |||
XM_017005224.1 | XP_016860713.1 | 21 | 2827 | |||
GenBank mRNA | AL133022.1 | - | 29 | 4746 | ||
AY461813.1 | - | 46 | 5369 | |||
BC117324.1 | - | 13 | 1551 | |||
BC126368.1 | - | 13 | 1554 | |||
BC143825.1 | - | 13 | 1551 |