Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TICRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308025.1 | NP_001294954.1 | 4 | 1354 | ||
NM_152259.3 | NP_689472.3 | 4 | 1357 | |||
GenBank mRNA | GQ227787.1 | - | 4 | 1354 |