Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080652.3 | NP_542383.1 | 4, 5 | 664 | ||
XM_017007437.1 | XP_016862926.1 | 3, 4 | 502 | |||
GenBank mRNA | AK290878.1 | - | 4, 5 | 642 | ||
AK313954.1 | - | 4, 5 | 600 | |||
AL832782.1 | - | 1, 2 | 123 | |||
AY358547.1 | - | 4, 5 | 648 | |||
BC008043.2 | - | 4, 5 | 621 | 3838280 | ||
BC019884.1 | - | 4, 5 | 611 | 4991416 | ||
BX647128.1 | - | 2, 3 | 314 |