Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021010.2 | NP_066290.1 | 1 | 203 | ||
GenBank mRNA | BC069690.1 | - | 1 | 199 | 7262486 | |
BC107079.2 | - | 1 | 194 | 40014027 | ||
BM854660.1 | - | 1 | 203 | |||
BP419449.1 | - | 1 | 148 |