Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319661.1 | NP_001306590.1 | 2 | 397 | ||
NM_001319662.1 | NP_001306591.1 | 2 | 418 | |||
NM_001319663.1 | NP_001306592.1 | 4 | 563 | |||
NM_001319664.1 | NP_001306593.1 | 4 | 581 | |||
NM_001319666.1 | NP_001306595.1 | 3 | 465 | |||
NM_152261.3 | NP_689474.1 | 3 | 465 | |||
XM_017020198.1 | XP_016875687.1 | 3 | 652 | |||
GenBank mRNA | AK001731.1 | - | 3 | 280 | ||
AK021783.1 | - | 1 | 80 | |||
AK022770.1 | - | 2 | 179 | |||
AK024021.1 | - | 3 | 292 | |||
BC020522.1 | - | 3 | 260 | 3916886 | ||
CX166555.1 | - | 4 | 457 | |||
DA609590.1 | - | 2 | 418 | |||
DB091654.1 | - | 3 | 465 | |||
DB226324.1 | - | 3 | 312 | |||
DB233011.1 | - | 4 | 386 | |||
DB458911.1 | - | 3 | 422 |