Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 6 | 680 | ||
NM_001039672.2 | NP_001034761.1 | 6 | 698 | |||
NM_001039673.2 | NP_001034762.1 | 6 | 689 | |||
NM_001145461.1 | NP_001138933.1 | 6 | 674 | |||
NM_001145462.1 | NP_001138934.1 | 6 | 650 | |||
NM_001145463.1 | NP_001138935.1 | 6 | 689 | |||
XM_005259385.3 | XP_005259442.1 | 6 | 911 | |||
XM_017027449.1 | XP_016882938.1 | 6 | 699 | |||
XM_017027450.1 | XP_016882939.1 | 6 | 697 | |||
XM_017027451.1 | XP_016882940.1 | 6 | 695 | |||
GenBank mRNA | AF258578.3 | - | 5 | 872 | ||
AK304479.1 | - | 3 | 1036 | |||
AL833382.1 | - | 6 | 646 | |||
AY358965.1 | - | 5 | 817 | |||
BC007644.1 | - | 6 | 637 | |||
BC014974.1 | - | 6 | 643 | 4865479 | ||
BC091477.1 | - | 6 | 672 | 30344388 | ||
BG765536.1 | - | 6 | 632 | |||
BX379501.2 | - | 6 | 616 | |||
CA487891.1 | - | 6 | 632 | |||
CA488303.1 | - | 6 | 627 | |||
CF127536.1 | - | 6 | 644 |