Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079691.1 | NP_001073159.1 | 2 | 387 | ||
NM_001286459.1 | NP_001273388.1 | 2 | 387 | |||
NM_001286460.1 | NP_001273389.1 | 2 | 387 | |||
NM_001286461.1 | NP_001273390.1 | 2 | 387 | |||
NM_052818.2 | NP_438169.2 | 2 | 387 | |||
XM_006719890.3 | XP_006719953.1 | 2 | 643 | |||
XM_011535301.2 | XP_011533603.1 | 3 | 540 | |||
XM_011535302.2 | XP_011533604.1 | 5 | 821 | |||
XM_011535303.2 | XP_011533605.1 | 2 | 950 | |||
XM_011535304.2 | XP_011533606.1 | 5 | 798 | |||
XM_017020836.1 | XP_016876325.1 | 2 | 1842 | |||
XM_017020837.1 | XP_016876326.1 | 2 | 627 | |||
XM_017020838.1 | XP_016876327.1 | 5 | 881 | |||
XM_017020840.1 | XP_016876329.1 | 5 | 796 | |||
XM_017020841.1 | XP_016876330.1 | 5 | 897 | |||
XM_017020842.1 | XP_016876331.1 | 5 | 821 | |||
GenBank mRNA | AI916535.1 | - | 2 | 223 | ||
AL049785.1 | - | 2 | 343 | |||
AL049786.1 | - | 2 | 343 | |||
AL049787.1 | - | 2 | 343 | |||
AL049788.1 | - | 4 | 401 | |||
BC022188.2 | - | 2 | 333 | |||
BC125050.1 | - | 1 | 56 | |||
BF448179.1 | - | 2 | 224 | |||
BI551105.1 | - | 2 | 392 | |||
BX648509.1 | - | 1 | 3316 | |||
U50527.1 | - | 2 | 338 |