Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031617.2 | NP_001026788.1 | 1, 2 | 173 | ||
GenBank mRNA | AK023185.1 | - | 1, 2 | 108 | ||
AY957566.1 | - | 1, 2 | 123 | |||
BC042434.1 | - | 1, 2 | 83 | |||
BC053372.1 | - | 1, 2 | 90 | |||
BC058158.1 | - | 1, 2 | 105 | |||
BC070383.1 | - | 1, 2 | 119 | 6724444 | ||
BC103632.1 | - | 1, 2 | 149 | 6606556 | ||
BC110420.1 | - | 1, 2 | 111 | 5208612 | ||
DB077585.1 | - | 1, 2 | 173 |
Gene Symbol: ADAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK290517.1 | - | 1, 2 | 123 |