Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF2LY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139214.2 | NP_631960.1 | 2 | 320 | ||
GenBank mRNA | AF332223.1 | - | 2 | 320 | ||
AK292609.1 | - | 2 | 364 | |||
BC128604.1 | - | 1 | 290 | 40003247 |
Gene Symbol: TGIF2LX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138960.3 | NP_620410.3 | 2 | 305 | ||
GenBank mRNA | AF497480.1 | - | 1 | 285 | ||
BC101144.2 | - | 1 | 290 | |||
BC101145.1 | - | 1 | 290 | |||
BC117398.1 | - | 2 | 305 | 40125949 | ||
BC117400.1 | - | 2 | 305 | 40125951 |