Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138360.3 | NP_612369.3 | 23 | 2070 | ||
XM_011537300.2 | XP_011535602.1 | 22 | 1729 | |||
XM_017021751.1 | XP_016877240.1 | 19 | 1630 | |||
GenBank mRNA | AK074167.1 | - | 10 | 817 |