Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRSAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005373.3 | NP_001005373.1 | 22 | 2037 | ||
NM_001005374.3 | NP_001005374.1 | 21 | 1742 | |||
NM_001190723.2 | NP_001177652.1 | 21 | 1956 | |||
NM_138361.5 | NP_612370.3 | 21 | 2296 | |||
XM_017015283.1 | XP_016870772.1 | 22 | 3036 | |||
XM_017015284.1 | XP_016870773.1 | 22 | 2032 | |||
GenBank mRNA | AK056203.1 | - | 21 | 1938 | ||
AK056305.1 | - | 10 | 887 | |||
AK091589.1 | - | 21 | 2044 | |||
AK097094.1 | - | 2 | 1497 | |||
AK226106.1 | - | 22 | 2023 | |||
AY358830.1 | - | 22 | 2002 | |||
BC009239.2 | - | 21 | 1742 | 2988664 |