Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138771.3 | NP_620126.2 | 3 | 587 | ||
XM_017012775.1 | XP_016868264.1 | 4 | 827 | |||
XM_017012776.1 | XP_016868265.1 | 3 | 570 | |||
GenBank mRNA | AK026684.1 | - | 1 | 84 | ||
AK289911.1 | - | 3 | 563 | |||
AY358713.1 | - | 3 | 554 | |||
BC012427.1 | - | 1 | 168 | 3858521 | ||
BC104638.1 | - | 3 | 479 | 6620343 |