Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138300.3 | NP_612157.1 | 1, 2 | 312 | ||
GenBank mRNA | AF289598.1 | - | 1, 2 | 64 | ||
AF457208.1 | - | 1, 2 | 106 | |||
AK092389.1 | - | 1, 2 | 261 | |||
BC006132.1 | - | 1, 2 | 278 | 3627860 | ||
BC032099.2 | - | 1, 2 | 278 | 4126360 | ||
CN260857.1 | - | 1, 2 | 315 |