Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316990.1 | NP_001303919.1 | 17 | 2410 | ||
NM_138363.2 | NP_612372.1 | 18 | 2516 | |||
NR_133644.1 | 18 | 2539 | ||||
XM_005257779.3 | XP_005257836.1 | 18 | 2492 | |||
XM_005257780.2 | XP_005257837.1 | 17 | 2135 | |||
XM_006722166.3 | XP_006722229.1 | 18 | 2274 | |||
XM_017025287.1 | XP_016880776.1 | 18 | 3495 | |||
XM_017025288.1 | XP_016880777.1 | 17 | 2168 | |||
GenBank mRNA | AK290587.1 | - | 18 | 2256 | ||
AK297409.1 | - | 17 | 2111 | |||
AL832822.1 | - | 18 | 2192 | |||
BC009518.1 | - | 18 | 2212 | 3633884 | ||
BX641136.1 | - | 1 | 2669 |