Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300753.1 | NP_001287682.1 | 9 | 1663 | ||
NM_001300755.1 | NP_001287684.1 | 9 | 1761 | |||
NM_001300757.1 | NP_001287686.1 | 8 | 1416 | |||
NM_001300758.1 | NP_001287687.1 | 8 | 1412 | |||
NM_144567.4 | NP_653168.2 | 9 | 1738 | |||
NR_125333.1 | 7 | 1285 | ||||
XM_005273344.1 | XP_005273401.1 | 9 | 1759 | |||
XM_005273345.1 | XP_005273402.1 | 9 | 1805 | |||
XM_005273346.2 | XP_005273403.1 | 9 | 1558 | |||
XM_005273347.3 | XP_005273404.1 | 8 | 1430 | |||
XM_017002774.1 | XP_016858263.1 | 9 | 1687 | |||
XM_017002775.1 | XP_016858264.1 | 10 | 1986 | |||
XM_017002776.1 | XP_016858265.1 | 10 | 2010 | |||
XM_017002777.1 | XP_016858266.1 | 10 | 2056 | |||
XM_017002778.1 | XP_016858267.1 | 10 | 1799 | |||
GenBank mRNA | AF510741.1 | - | 8 | 1367 | ||
AK093947.1 | - | 4 | 2162 | |||
AK294479.1 | - | 9 | 1537 | |||
AK295116.1 | - | 9 | 1667 | |||
AK299208.1 | - | 9 | 1731 | |||
AK299365.1 | - | 7 | 1195 | |||
AK310946.1 | - | 7 | 904 | |||
AL079275.1 | - | 9 | 1696 | |||
BC047469.1 | - | 8 | 1328 | 5274835 |