Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001927.3 | NP_001918.3 | 8 | 1440 | ||
GenBank mRNA | AF055081.1 | - | 8 | 1354 | ||
AF055082.1 | - | 8 | 1354 | |||
AF055083.1 | - | 8 | 1354 | |||
AF167579.1 | - | 8 | 1354 | |||
AF486807.1 | - | 8 | 1354 | |||
AF487828.1 | - | 8 | 1354 | |||
AF521879.1 | - | 8 | 1351 | |||
AF527578.1 | - | 8 | 1354 | |||
AK022087.1 | - | 1630 | ||||
AK097038.1 | - | 9 | 1380 | |||
AK098332.1 | - | 9 | 1234 | |||
AK123787.1 | - | 9 | 1179 | |||
AK300654.1 | - | 1158 | ||||
AL541778.3 | - | 6 | 716 | |||
AY083345.1 | - | 8 | 1354 | |||
AY125465.1 | - | 8 | 1354 | |||
BC010072.2 | - | 8 | 1424 | |||
BC032116.2 | - | 8 | 1452 | 4905678 | ||
DQ104335.1 | - | 8 | 1354 | |||
DQ104336.1 | - | 8 | 1354 | |||
DQ104337.1 | - | 8 | 1354 | |||
EF617312.1 | - | 8 | 1434 | |||
JX114780.1 | - | 8 | 1354 | |||
U59167.1 | - | 8 | 1431 |