Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 10 | 2289 | ||
NM_001281988.1 | NP_001268917.1 | 9 | 867 | |||
NM_144569.5 | NP_653170.3 | 10 | 2289 | |||
XM_011542413.2 | XP_011540715.1 | 10 | 858 | |||
XM_011542414.2 | XP_011540716.1 | 9 | 1053 | |||
XM_017002779.1 | XP_016858268.1 | 9 | 2198 | |||
XM_017002780.1 | XP_016858269.1 | 8 | 2156 | |||
XM_017002782.1 | XP_016858271.1 | 9 | 1427 | |||
GenBank mRNA | AK091016.1 | - | 9 | 838 | ||
AK095829.1 | - | 3 | 465 | |||
AK096192.1 | - | 2 | 439 | |||
AK097227.1 | - | 7 | 693 | |||
AK131447.1 | - | 10 | 2218 | |||
BC007910.2 | - | 3 | 355 | |||
BC114430.1 | - | 10 | 2215 | 40038407 |