Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 11 | 2478 | ||
NM_001281988.1 | NP_001268917.1 | 10 | 1056 | |||
NM_144569.5 | NP_653170.3 | 11 | 2478 | |||
XM_011542413.2 | XP_011540715.1 | 11 | 1047 | |||
XM_011542414.2 | XP_011540716.1 | 10 | 1242 | |||
XM_017002779.1 | XP_016858268.1 | 10 | 2384 | |||
XM_017002780.1 | XP_016858269.1 | 9 | 2345 | |||
XM_017002781.1 | XP_016858270.1 | 9 | 2261 | |||
XM_017002782.1 | XP_016858271.1 | 10 | 1616 | |||
GenBank mRNA | AK058077.1 | - | 1 | 58 | ||
AK091016.1 | - | 10 | 1027 | |||
AK131447.1 | - | 11 | 2407 | |||
BC007910.2 | - | 4 | 541 | |||
BC029069.1 | - | 3 | 368 | |||
BC114430.1 | - | 11 | 2404 | 40038407 |