Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318058.1 | NP_001304987.1 | 2 | 1070 | ||
NM_001318059.1 | NP_001304988.1 | 2 | 972 | |||
NM_033428.2 | NP_219500.1 | 2 | 1058 | |||
GenBank mRNA | AL137489.1 | - | 2 | 862 |