Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138961.2 | NP_620411.2 | 5 | 801 | ||
XM_005271720.4 | XP_005271777.1 | 2 | 222 | |||
XM_011543060.2 | XP_011541362.1 | 4 | 564 | |||
GenBank mRNA | AF361746.1 | - | 5 | 766 | ||
AK075396.1 | - | 5 | 764 | |||
AK091023.1 | - | 5 | 1684 | |||
AK092429.1 | - | 3 | 1173 | |||
AK301160.1 | - | 2 | 227 | |||
AK314710.1 | - | 5 | 764 | |||
AY189281.1 | - | 2 | 1347 | |||
AY358382.1 | - | 5 | 753 | |||
BC016868.1 | - | 5 | 635 | 3850665 |