Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138961.2 | NP_620411.2 | 3 | 561 | ||
XM_011543060.2 | XP_011541362.1 | 2 | 324 | |||
GenBank mRNA | AF361746.1 | - | 3 | 526 | ||
AK075396.1 | - | 3 | 524 | |||
AK091023.1 | - | 3 | 1476 | |||
AK092429.1 | - | 1 | 933 | |||
AK314710.1 | - | 3 | 524 | |||
AY189281.1 | - | 1 | 927 | |||
AY358382.1 | - | 3 | 513 | |||
BC016868.1 | - | 3 | 395 | 3850665 |