Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052853.3 | NP_443085.2 | 4 | 1430 | ||
XM_006716170.3 | XP_006716233.1 | 4 | 1655 | |||
GenBank mRNA | AF131745.1 | - | 4 | 1429 | ||
AY302592.1 | - | 4 | 1430 |