Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052853.3 | NP_443085.2 | 2, 3 | 1266 | ||
XM_006716170.3 | XP_006716233.1 | 2, 3 | 1491 | |||
XM_011516675.2 | XP_011514977.1 | 2, 3 | 1491 | |||
GenBank mRNA | AF131745.1 | - | 2, 3 | 1265 | ||
AY302592.1 | - | 2, 3 | 1266 | |||
BC014107.2 | - | 2, 3 | 1263 | 4138734 |