Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198963.1 | NP_945314.1 | 14, 15 | 2732 | ||
XM_011533154.2 | XP_011531456.1 | 14, 15 | 2736 | |||
XM_011533155.2 | XP_011531457.1 | 14, 15 | 2734 | |||
XM_011533156.2 | XP_011531458.1 | 14, 15 | 2736 | |||
XM_011533157.1 | XP_011531459.1 | 14, 15 | 2736 | |||
GenBank mRNA | AB209249.1 | - | 9, 10 | 1219 | ||
AF283512.1 | - | 10, 11 | 1423 | |||
AK296739.1 | - | 14, 15 | 2771 | |||
AK307514.1 | - | 14, 15 | 2761 | |||
BC039270.1 | - | 13, 14 | 2562 | |||
BC053623.1 | - | 8, 9 | 1033 | |||
BC065278.1 | - | 9, 10 | 1227 | |||
BC131534.1 | - | 14, 15 | 2704 | 40082400 |