Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317335.1 | NP_001304264.1 | 5 | 947 | ||
NM_001928.3 | NP_001919.2 | 5 | 926 | |||
GenBank mRNA | AK300963.1 | - | 5 | 921 | ||
BC034529.1 | - | 4 | 842 | |||
BC051001.1 | - | 5 | 866 | |||
BI763909.1 | - | 302 | ||||
M84526.1 | - | 5 | 874 |