Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175736.4 | NP_783863.4 | 23, 24 | 3005 | ||
NM_198900.2 | NP_944489.2 | 22, 23 | 2852 | |||
XM_005269218.2 | XP_005269275.1 | 23, 24 | 2984 | |||
XM_011538968.2 | XP_011537270.1 | 23, 24 | 3474 | |||
XM_011538969.2 | XP_011537271.1 | 23, 24 | 3473 | |||
XM_011538970.2 | XP_011537272.1 | 23, 24 | 3473 | |||
XM_011538971.2 | XP_011537273.1 | 23, 24 | 3049 | |||
XM_011538972.2 | XP_011537274.1 | 22, 23 | 3319 | |||
XM_011538973.2 | XP_011537275.2 | 19, 20 | 2464 | |||
XM_011538974.2 | XP_011537276.1 | 24, 25 | 2806 | |||
GenBank mRNA | AB095934.2 | - | 23, 24 | 3005 | ||
AK127200.1 | - | 10, 11 | 1478 | |||
AK128195.1 | - | 22, 23 | 2832 | |||
AL162062.1 | - | 7, 8 | 962 | |||
BC033181.1 | - | 7, 8 | 789 | |||
BC159100.1 | - | 23, 24 | 3005 | 8860524 |