Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281731.1 | NP_001268660.1 | 4 | 790 | ||
NM_147190.3 | NP_671723.1 | 3 | 589 | |||
NR_104035.1 | 3 | 589 | ||||
XM_005269220.2 | XP_005269277.1 | 3 | 425 | |||
XM_006719687.3 | XP_006719750.1 | 3 | 425 | |||
XM_011538975.2 | XP_011537277.1 | 3 | 425 | |||
XM_017020202.1 | XP_016875691.1 | 3 | 349 | |||
XM_017020203.1 | XP_016875692.1 | 4 | 713 | |||
XM_017020204.1 | XP_016875693.1 | 3 | 425 | |||
XM_017020205.1 | XP_016875694.1 | 3 | 278 | |||
GenBank mRNA | AK026014.1 | - | 3 | 395 | ||
AK058033.1 | - | 1 | 981 | |||
AK294521.1 | - | 3 | 363 | |||
AK298924.1 | - | 2 | 343 | |||
AK300937.1 | - | 4 | 565 | |||
BC032565.1 | - | 3 | 406 | 5580489 | ||
BC033558.1 | - | 3 | 376 | |||
U15426.1 | - | 456 |