Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052997.2 | NP_443723.2 | 34 | 3982 | ||
XM_011519757.2 | XP_011518059.1 | 42 | 4526 | |||
XM_017016883.1 | XP_016872372.1 | 22 | 2355 | |||
GenBank mRNA | AF269087.1 | - | 34 | 3982 |