Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164586.1 | NP_001158058.1 | 21 | 10484 | ||
XM_005245580.3 | XP_005245637.1 | 21 | 10454 | |||
XM_011510120.2 | XP_011508422.1 | 21 | 10576 | |||
XM_011510121.2 | XP_011508423.1 | 21 | 10561 | |||
XM_017002787.1 | XP_016858276.1 | 20 | 10256 | |||
GenBank mRNA | AY245430.2 | - | 10 | 2606 | ||
BC033185.1 | - | 23 | 3063 | |||
BC131529.1 | - | 10 | 2187 | 40036345 |