Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198805.1 | NP_001185734.1 | 2 | 299 | ||
NM_001198806.1 | NP_001185735.1 | 2 | 306 | |||
NM_001198807.1 | NP_001185736.1 | 2 | 338 | |||
NM_080655.2 | NP_542386.1 | 2 | 338 | |||
GenBank mRNA | AK314924.1 | - | 2 | 198 | ||
AY598327.1 | - | 1 | 124 | |||
BC008993.1 | - | 2 | 191 | 4213591 | ||
BI596498.1 | - | 2 | 296 | |||
BM914752.1 | - | 2 | 217 | |||
CN366267.1 | - | 2 | 296 | |||
DA424604.1 | - | 2 | 306 | |||
DR159535.1 | - | 2 | 338 |
Gene Symbol: MSANTD3-TMEFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198812.1 | NP_001185741.1 | 1 | 157 |