Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 7 | 1248 | ||
NM_001320132.1 | NP_001307061.1 | 7 | 1345 | |||
NM_001320136.1 | NP_001307065.1 | 7 | 1226 | |||
NM_004402.3 | NP_004393.1 | 7 | 1176 | |||
NR_104222.1 | 8 | 1313 | ||||
NR_135150.1 | 8 | 1300 | ||||
NR_135151.1 | 8 | 1263 | ||||
NR_135152.1 | 7 | 1198 | ||||
XM_017000498.1 | XP_016855987.1 | 6 | 1111 | |||
XM_017000499.1 | XP_016855988.1 | 6 | 1127 | |||
XM_017000500.1 | XP_016855989.1 | 6 | 882 | |||
GenBank mRNA | AB013918.1 | - | 7 | 839 | ||
AB028911.1 | - | 8 | 1093 | |||
AB028912.1 | - | 7 | 1139 | |||
AB028913.1 | - | 8 | 1057 | |||
AF039210.1 | - | 7 | 839 | |||
AF064019.1 | - | 7 | 970 | |||
AF409060.1 | - | 9 | 850 | |||
AF409061.1 | - | 7 | 865 | |||
AF409062.1 | - | 7 | 893 | |||
AK290877.1 | - | 7 | 967 | |||
AK303065.1 | - | 7 | 1055 | |||
AY438267.1 | - | 7 | 768 | |||
AY438268.1 | - | 4 | 391 | |||
BC032827.1 | - | 8 | 1125 | |||
BC048797.1 | - | 7 | 1162 | 5204996 |