Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012967.2 | NP_001012985.2 | 27 | 3858 | ||
NM_001291510.1 | NP_001278439.1 | 27 | 3861 | |||
XM_005263341.4 | XP_005263398.2 | 27 | 3844 | |||
XM_005263342.2 | XP_005263399.2 | 27 | 3861 | |||
XM_005263343.2 | XP_005263400.2 | 27 | 3827 | |||
XM_005263345.4 | XP_005263402.2 | 27 | 3840 | |||
XM_006714408.2 | XP_006714471.1 | 28 | 3911 | |||
XM_011532402.2 | XP_011530704.1 | 28 | 4134 | |||
XM_011532403.2 | XP_011530705.1 | 28 | 3969 | |||
XM_011532404.1 | XP_011530706.1 | 26 | 3765 | |||
XM_011532405.1 | XP_011530707.1 | 26 | 3762 | |||
XM_011532406.1 | XP_011530708.1 | 27 | 3861 | |||
XM_011532409.2 | XP_011530711.1 | 15 | 1901 | |||
XM_017008829.1 | XP_016864318.1 | 26 | 3731 | |||
XM_017008830.1 | XP_016864319.1 | 27 | 3861 | |||
XM_017008831.1 | XP_016864320.1 | 16 | 2163 | |||
GenBank mRNA | AK023743.1 | - | 6 | 744 | ||
AL832950.1 | - | 8 | 953 | |||
CR933601.1 | - | 27 | 3836 | |||
KR632542.1 | - | 25 | 3610 |