Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 6, 7 | 1207 | ||
XM_006717317.3 | XP_006717380.1 | 6, 7 | 1233 | |||
XM_011519182.2 | XP_011517484.1 | 5, 6 | 801 | |||
GenBank mRNA | AK022632.1 | - | 1, 2 | 92 | ||
AK095393.1 | - | 6, 7 | 948 | |||
AK128529.1 | - | 4, 5 | 1042 | |||
AL832421.1 | - | 6, 7 | 976 | |||
BC106035.1 | - | 6, 7 | 919 | |||
BC136391.1 | - | 5, 6 | 774 |