Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 6 | 1163 | ||
XM_006717317.3 | XP_006717380.1 | 6 | 1189 | |||
XM_011519182.2 | XP_011517484.1 | 5 | 757 | |||
GenBank mRNA | AK022632.1 | - | 1 | 48 | ||
AK074143.1 | - | 3 | 1092 | |||
AK095393.1 | - | 6 | 904 | |||
AK128529.1 | - | 4 | 998 | |||
AL832421.1 | - | 6 | 932 | |||
BC106035.1 | - | 6 | 875 | |||
BC136391.1 | - | 5 | 730 |