Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136497.2 | NP_001129969.1 | 2 | 263 | ||
NM_001243767.1 | NP_001230696.1 | 3 | 330 | |||
NM_152265.4 | NP_689478.1 | 3 | 330 | |||
GenBank mRNA | AK027582.1 | - | 2 | 144 | ||
AK027750.1 | - | 3 | 212 | |||
BC021004.1 | - | 2 | 107 | |||
BC022371.1 | - | 3 | 192 | 4739327 | ||
BC070378.1 | - | 3 | 220 | 6495112 | ||
BU567198.1 | - | 3 | 333 | |||
BU785869.1 | - | 3 | 174 | |||
DA369498.1 | - | 3 | 251 |