Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018392.4 | NP_060862.3 | 19 | 5024 | ||
XM_005263115.3 | XP_005263172.1 | 19 | 5079 | |||
XM_011532091.2 | XP_011530393.1 | 18 | 4973 | |||
XM_011532092.2 | XP_011530394.1 | 18 | 4904 | |||
XM_011532093.2 | XP_011530395.1 | 18 | 4850 | |||
XM_011532094.2 | XP_011530396.1 | 17 | 4766 | |||
XM_011532097.2 | XP_011530399.1 | 12 | 2126 | |||
XM_011532098.2 | XP_011530400.1 | 10 | 1800 | |||
XM_011532099.2 | XP_011530401.1 | 9 | 1759 | |||
XM_017008369.1 | XP_016863858.1 | 18 | 5019 | |||
XM_017008370.1 | XP_016863859.1 | 17 | 4799 | |||
XM_017008371.1 | XP_016863860.1 | 16 | 4679 | |||
XM_017008372.1 | XP_016863861.1 | 16 | 4058 | |||
XM_017008373.1 | XP_016863862.1 | 15 | 3995 | |||
GenBank mRNA | AK090556.1 | - | 1 | 659 | ||
AK126054.1 | - | 8 | 1408 | |||
AL137700.1 | - | 5 | 599 | |||
BX647594.1 | - | 8 | 1507 |