Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152266.4 | NP_689479.1 | 3 | 285 | ||
GenBank mRNA | AK128668.1 | - | 3 | 252 | ||
BC003535.1 | - | 3 | 205 | 3609326 | ||
BC010170.2 | - | 3 | 213 | 3659284 | ||
BC020247.1 | - | 3 | 233 | 4620233 | ||
DA006539.1 | - | 3 | 260 | |||
KU178830.1 | - | 2 | 142 |