Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173465.3 | NP_775736.2 | 16 | 1283 | ||
XM_006714933.3 | XP_006714996.1 | 16 | 1013 | |||
XM_006714934.3 | XP_006714997.1 | 15 | 986 | |||
XM_006714935.3 | XP_006714998.1 | 15 | 977 | |||
XM_006714936.3 | XP_006714999.1 | 15 | 950 | |||
XM_011534688.2 | XP_011532990.1 | 15 | 986 | |||
XM_011534689.2 | XP_011532991.1 | 16 | 1013 | |||
XM_011534690.2 | XP_011532992.1 | 14 | 959 | |||
XM_011534691.2 | XP_011532993.1 | 12 | 905 | |||
XM_011534692.2 | XP_011532994.1 | 16 | 2038 | |||
GenBank mRNA | AY158895.1 | - | 16 | 1283 | ||
AY898961.1 | - | 16 | 1283 | |||
BC042428.1 | - | 17 | 1065 | 5165083 |